CH6371381098 Chem Impex International, Inc.

Fmoc-S-trityl-D-homocysteine

  • MDL number MFCD05663440
  • Cas Number : 1007840-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Homocys(Trt)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-s-trityl-d-homocysteine/6308?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-s-trityl-d-homocysteine/6308?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart