CH6371381103 Chem Impex International, Inc.

2-Iodo-D-phenylalanine

  • MDL number MFCD08458534
  • Cas Number : 736184-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201 - 207 �dec.)
Synonym D-Phe(2-I)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-iodo-d-phenylalanine/6313?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-iodo-d-phenylalanine/6313?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart