CH6371381106 Chem Impex International, Inc.

Na,Na-Bis-Fmoc-L-cystine bis(tert-butyl ester)

  • MDL number MFCD08458535
  • Cas Number : 139592-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Fmoc-L-Cys-OtBu)2
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-n-i-sup-alpha-sup-bis-fmoc-l-cystine-bis-i-tert-i-butyl-ester/6317?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-n-i-sup-alpha-sup-bis-fmoc-l-cystine-bis-i-tert-i-butyl-ester/6317?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart