CH6371381111 Chem Impex International, Inc.

Na-Ac-Ne-2-Fmoc-L-lysine

  • MDL number MFCD08458546
  • Cas Number : 148101-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-169
Synonym Ac-L-Lys(Fmoc)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-ac-i-n-i-sup-epsilon-sup-2-fmoc-l-lysine/6340?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-ac-i-n-i-sup-epsilon-sup-2-fmoc-l-lysine/6340?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart