CH6371381115 Chem Impex International, Inc.

Cyanoacetic acid N-hydroxysuccinimide ester

  • MDL number MFCD00798802
  • Cas Number : 56657-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Sotre at = -4
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyanoacetic-acid-n-hydroxysuccinimide-ester/6350?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyanoacetic-acid-n-hydroxysuccinimide-ester/6350?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart