CH6371381122 Chem Impex International, Inc.

Na-Fmoc-Nw-bis-carbobenzoxy-L-arginine

  • MDL number MFCD05663439
  • Cas Number : 207857-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Arg(Z)2-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-w-sup-bis-carbobenzoxy-l-arginine/6366?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-w-sup-bis-carbobenzoxy-l-arginine/6366?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart