CH6371381124 Chem Impex International, Inc.

(S)-3-(4-Boc-piperazin-1-yl)-2-(Fmoc-amino)propionic acid

  • MDL number MFCD01860747
  • Cas Number : 313052-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-4-boc-piperazin-1-yl-2-fmoc-amino-propionic-acid/6371?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-4-boc-piperazin-1-yl-2-fmoc-amino-propionic-acid/6371?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart