CH6371381129 Chem Impex International, Inc.

Fmoc-D-leucinol

  • MDL number MFCD00270276
  • Cas Number : 215178-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-139 �C
Synonym Fmoc-D-Leu-ol
Purity 99.5
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-leucinol/6382?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-leucinol/6382?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart