CH6371381130 Chem Impex International, Inc.

Na-Fmoc-Nw-bis-carbobenzoxy-L-homoarginine

  • MDL number MFCD08458579
  • Cas Number : 1926163-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Homoarg(Z)2-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-omega-sup-bis-carbobenzoxy-l-homoarginine/6383?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-omega-sup-bis-carbobenzoxy-l-homoarginine/6383?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart