CH6371381131 Chem Impex International, Inc.

Fmoc-L-homoarginine

  • MDL number MFCD01861360
  • Cas Number : 776277-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Homoarg-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-homoarginine/6384?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-homoarginine/6384?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart