CH6371381133 Chem Impex International, Inc.

Na-Fmoc-Ne-7-methoxycoumarin-4-acetyl-L-lysine

  • MDL number MFCD03791024
  • Cas Number : 386213-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-206� C
Synonym Fmoc-L-Lys(Mca)-OH
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-7-methoxycoumarin-4-acetyl-l-lysine/6387?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-7-methoxycoumarin-4-acetyl-l-lysine/6387?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart