CH6371381136 Chem Impex International, Inc.

Na-Fmoc-Nin-formyl-L-tryptophan

  • MDL number MFCD08458583
  • Cas Number : 152338-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-194
Synonym Fmoc-L-Trp(For)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-in-sup-formyl-l-tryptophan/6392?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-in-sup-formyl-l-tryptophan/6392?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart