CH6371381140 Chem Impex International, Inc.

a-Aminoisobutyric acid methyl ester hydrochloride

  • MDL number MFCD00214247
  • Cas Number : 15028-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-182 �C
Synonym Aib-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/alpha-aminoisobutyric-acid-methyl-ester-hydrochloride/6396?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/alpha-aminoisobutyric-acid-methyl-ester-hydrochloride/6396?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart