CH6371381143 Chem Impex International, Inc.

3-(3-Pyridyl)-D-alanine methyl ester dihydrochloride

  • MDL number MFCD01317722
  • Cas Number : 197088-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym beta-(3-Pyridyl)-D-Ala-OMe 2HCl
Purity 0.97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-3-pyridyl-d-alanine-methyl-ester-dihydrochloride/6400?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-3-pyridyl-d-alanine-methyl-ester-dihydrochloride/6400?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart