CH6371381144 Chem Impex International, Inc.

beta-Ala-pNA HBr

  • MDL number MFCD08458595
  • Cas Number : 111196-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-206° C
Purity 99
Storage temp. Store at RT
Physical Form Slightly yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ß-ala-pna·hbr/6401?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-ala-pna·hbr/6401?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart