CH6371381145 Chem Impex International, Inc.

D-2-Aminobutyric acid tert-butyl ester hydrochloride

  • MDL number MFCD08275809
  • Cas Number : 313994-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-2-aminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/6402?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-2-aminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/6402?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart