CH6371381149 Chem Impex International, Inc.

D-alpha,beta-Diaminopropionic acid(Boc) methyl ester hydrochloride

  • MDL number MFCD05663450
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-137 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-alpha-beta-diaminopropionic-acid-boc-methyl-ester-hydrochloride/6408?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-alpha-beta-diaminopropionic-acid-boc-methyl-ester-hydrochloride/6408?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart