CH6371381154 Chem Impex International, Inc.

g-Aminobutyric acid benzyl ester p-tosylate

  • MDL number MFCD03424226
  • Cas Number : 26727-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym gamma-Abu-OBzl TosOH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/aminobutyric-acid-benzyl-ester-i-p-i-tosylate/6414?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/aminobutyric-acid-benzyl-ester-i-p-i-tosylate/6414?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart