CH6371381158 Chem Impex International, Inc.

Boc-5-amino-2-methoxybenzoic acid

  • MDL number MFCD04972601
  • Cas Number : 1075242-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-171 � C
Purity 99
Storage temp. Store at 0-8
Physical Form White amorphous powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-5-amino-2-methoxybenzoic-acid/6418?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-5-amino-2-methoxybenzoic-acid/6418?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart