CH6371381169 Chem Impex International, Inc.

Boc-(S)-3-amino-2-(phenylsulfonylamino)propionic acid

  • MDL number MFCD04112668
  • Cas Number : 342888-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-140
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-2-phenylsulfonylamino-propionic-acid/6430?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-2-phenylsulfonylamino-propionic-acid/6430?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart