CH6371381170 Chem Impex International, Inc.

Boc-(S)-3-amino-2-(p-toluenesulfonylamino)propionic acid

  • MDL number MFCD04112669
  • Cas Number : 16947-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-126
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-2-p-toluenesulfonylamino-propionic-acid/6431?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-2-p-toluenesulfonylamino-propionic-acid/6431?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart