CH6371381172 Chem Impex International, Inc.

Boc-3-amino-5-(Fmoc-amino)benzoic acid

  • MDL number MFCD04112671
  • Cas Number : 779335-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-183
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-amino-5-fmoc-amino-benzoic-acid/6433?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-amino-5-fmoc-amino-benzoic-acid/6433?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart