CH6371381174 Chem Impex International, Inc.

N-(1-Boc-piperidin-4-yl)-L-aspartic acid-4-benzyl ester

  • MDL number MFCD04112673
  • Cas Number : 959584-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White amorphous powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-1-boc-piperidin-4-yl-l-aspartic-acid-4-benzyl-ester/6435?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-1-boc-piperidin-4-yl-l-aspartic-acid-4-benzyl-ester/6435?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart