CH6371381177 Chem Impex International, Inc.

Boc-3-amino-2-naphthoic acid

  • MDL number MFCD02682309
  • Cas Number : 887242-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-189 °C
Purity 97
Storage temp. Store at 0-8°C
Physical Form White to yellowish-grey powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-amino-2-naphthoic-acid/6438?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-amino-2-naphthoic-acid/6438?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart