CH6371381180 Chem Impex International, Inc.

Boc-(S)-2-amino-3-(3-indolyl)propionitrile

  • MDL number MFCD04112675
  • Cas Number : 138165-79-4
  • Molecular Weight : 285.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-2-amino-3-3-indolyl-propionitrile/6441?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-2-amino-3-3-indolyl-propionitrile/6441?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart