CH6371381185 Chem Impex International, Inc.

3-(Boc-aminomethyl)benzylamine hydrochloride

  • MDL number MFCD04112678
  • Cas Number : 914465-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-195 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-boc-aminomethyl-benzylamine-hydrochloride/6446?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-boc-aminomethyl-benzylamine-hydrochloride/6446?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart