CH6371381197 Chem Impex International, Inc.

Fmoc-3-pyrenyl-L-alanine

  • MDL number MFCD01861376
  • Cas Number : 183071-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Ala(pyrenyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-pyrenyl-l-alanine/6458?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-pyrenyl-l-alanine/6458?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart