CH6371381198 Chem Impex International, Inc.

Fmoc-N-methyl-2-aminobenzoic acid

  • MDL number MFCD04972602
  • Cas Number : 120467-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-142 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-2-aminobenzoic-acid/6459?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-2-aminobenzoic-acid/6459?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart