CH6371381203 Chem Impex International, Inc.

Fmoc-(S)-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD02682321
  • Cas Number : 141636-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-73 �C
Synonym Fmoc-(S)-Dtc-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-5-5-dimethyl-1-3-thiazolidine-4-carboxylic-acid/6464?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-5-5-dimethyl-1-3-thiazolidine-4-carboxylic-acid/6464?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart