CH6371381213 Chem Impex International, Inc.

Fmoc-(R,S)-3-amino-3-(biphenyl)propionic acid

  • MDL number MFCD02179646
  • Cas Number : 269078-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-DL-3-(4-diphenyl)-3-amino-propionic acid
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-3-amino-3-biphenyl-propionic-acid/6474?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-3-amino-3-biphenyl-propionic-acid/6474?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart