CH6371381215 Chem Impex International, Inc.

Fmoc-(R,S)-3-amino-3-(2-naphthyl)propionic acid

  • MDL number MFCD01863204
  • Cas Number : 269078-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-3-amino-3-2-naphthyl-propionic-acid/6476?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-3-amino-3-2-naphthyl-propionic-acid/6476?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart