CH6371381221 Chem Impex International, Inc.

Fmoc-(S)3-amino-2-(phenylsulfonylamino)propionic acid

  • MDL number MFCD04112690
  • Cas Number : 273399-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-220
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-2-phenylsulfonylamino-propionic-acid/6482?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-2-phenylsulfonylamino-propionic-acid/6482?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart