CH6371381227 Chem Impex International, Inc.

Fmoc-beta-(benzothiazol-2-yl)-L-alanine

  • MDL number MFCD03791021
  • Cas Number : 959583-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99.5
Storage temp. Store at 0-8 °C
Physical Form White or off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-beta-benzothiazol-2-yl-l-alanine/6488?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-beta-benzothiazol-2-yl-l-alanine/6488?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart