CH6371381247 Chem Impex International, Inc.

Boc-3-bromo-D-phenylalanine

  • MDL number MFCD01317704
  • Cas Number : 261360-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Phe(3-Br)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-bromo-d-phenylalanine/6509?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-bromo-d-phenylalanine/6509?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart