CH6371381248 Chem Impex International, Inc.

Boc-3-chloro-D-tyrosine dicyclohexylamine salt

  • MDL number MFCD19690977

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-m-chloro-D-Tyr-OH DCHA
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-chloro-d-tyrosine-dicyclohexylamine-salt/6510?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-chloro-d-tyrosine-dicyclohexylamine-salt/6510?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart