CH6371381249 Chem Impex International, Inc.

Boc-3-iodo-D-phenylalanine

  • MDL number MFCD03840394
  • Cas Number : 478183-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-121 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-iodo-d-phenylalanine/6511?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-iodo-d-phenylalanine/6511?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart