CH6371381252 Chem Impex International, Inc.

Boc-2-iodo-L-phenylalanine

  • MDL number MFCD01860645
  • Cas Number : 273221-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-163 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-iodo-l-phenylalanine/6514?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-iodo-l-phenylalanine/6514?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart