CH6371381260 Chem Impex International, Inc.

Fmoc-2-iodo-D-phenylalanine

  • MDL number MFCD06796745
  • Cas Number : 478183-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-138
Synonym Fmoc-D-Phe(2-I)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-iodo-d-phenylalanine/6522?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-iodo-d-phenylalanine/6522?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart