CH6371381261 Chem Impex International, Inc.

Fmoc-2-nitro-D-phenylalanine

  • MDL number MFCD01317714
  • Cas Number : 478183-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Phe(2-NO2)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-nitro-d-phenylalanine/6523?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-nitro-d-phenylalanine/6523?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart