CH6371381263 Chem Impex International, Inc.

Fmoc-3-chloro-D-tyrosine

  • MDL number MFCD03840399
  • Cas Number : 478183-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-m-chloro-D-Tyr-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-chloro-d-tyrosine/6525?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-chloro-d-tyrosine/6525?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart