CH6371381265 Chem Impex International, Inc.

Fmoc-3-iodo-D-tyrosine

  • MDL number MFCD03840398
  • Cas Number : 244028-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 157 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-iodo-d-tyrosine/6527?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-iodo-d-tyrosine/6527?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart