CH6371381289 Chem Impex International, Inc.

Cefepime HCl

  • MDL number MFCD06795759
  • Cas Number : 123171-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 8490
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cefepime·hcl/6555?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cefepime·hcl/6555?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart