CH6371381295 Chem Impex International, Inc.

Boc-N-methyl-DL-phenylglycine

  • MDL number MFCD06796751
  • Cas Number : 30925-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-121 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-dl-phenylglycine/6563?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-dl-phenylglycine/6563?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart