CH6371381299 Chem Impex International, Inc.

Dicloxacillin Sodium

  • MDL number MFCD00056865
  • Cas Number : 13412-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-225
Purity 95102
Storage temp. Store at 0-8
Physical Form White to almost white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dicloxacillin-sodium-usp-bp/6567?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dicloxacillin-sodium-usp-bp/6567?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart