CH6371381310 Chem Impex International, Inc.

Boc-4-nitro-L-beta-homophenylalanine

  • MDL number MFCD01076286
  • Cas Number : 127106-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-nitro-l-beta-homophenylalanine/6579?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-nitro-l-beta-homophenylalanine/6579?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart