CH6371381311 Chem Impex International, Inc.

(S)-3-Amino-3-(3-pyridyl)propionic acid

  • MDL number MFCD04113694
  • Cas Number : 129043-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-217 �dec.)
Synonym L-beta-Ala-(3-pyridyl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-pyridyl-propionic-acid/6581?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-pyridyl-propionic-acid/6581?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart