CH6371381313 Chem Impex International, Inc.

(R)-3-Amino-3-(4-chlorophenyl)propionic acid

  • MDL number MFCD03840387
  • Cas Number : 131690-61-4
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-chlorophenyl-propionic-acid/6585?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-chlorophenyl-propionic-acid/6585?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart