CH6371381314 Chem Impex International, Inc.

(S)-3-Amino-3-(2-furyl)propionic acid

  • MDL number MFCD04117828
  • Cas Number : 131829-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-222 �C (dec.)
Synonym D-beta-Ala-(2-furyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-furyl-propionic-acid/6586?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-furyl-propionic-acid/6586?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart