CH6371381315 Chem Impex International, Inc.

(+)-(1R,4S)-4-Aminocyclopent-2-enecarboxylic acid

  • MDL number MFCD00211285
  • Cas Number : 134003-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (+)-(1R,4S)-gamma-Homocycloleu-2-ene
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-r-i-4-i-s-i-4-aminocyclopent-2-enecarboxylic-acid/6587?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-r-i-4-i-s-i-4-aminocyclopent-2-enecarboxylic-acid/6587?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart