CH6371381320 Chem Impex International, Inc.

(R)-3-Amino-3-(2-fluorophenyl)propionic acid

  • MDL number MFCD04113643
  • Cas Number : 151911-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-fluorophenyl-propionic-acid/6592?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-fluorophenyl-propionic-acid/6592?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart